Merck
  • [1-butanol synthesis by Escherichia coli cells through butyryl-CoA formation by heterologous enzymes of clostridia and native enzymes of fatty acid beta-oxidation].
[1-butanol synthesis by Escherichia coli cells through butyryl-CoA formation by heterologous enzymes of clostridia and native enzymes of fatty acid beta-oxidation].
Prikladnaia biokhimiia i mikrobiologiia (2012-10-06)
A Iu Gulevich, A Iu Skorokhodova, A A Morzhakova, S V Antonova, A V Sukhozhenko, R S Shakulov, V G Debabov
ABSTRACT
Anaerobic biosynthesis of 1-butanol from glucose is investigated in recombinant Escherichia coli strains which form butyryl-CoA using the heterologous enzyme complex of clostridia or as a result of a reversal in the action of native enzymes of the fatty acid beta-oxidation pathway. It was revealed that when the basic pathways of acetic and lactic acid formation are inactivated due to deletions in the ackA, pta, poxB, and ldhA genes, the efficiency of butyryl-CoA biosynthesis and its reduced product, i.e., 1-butanol, by two types of recombinant stains is comparable. The limiting factor for 1-butanol production by the obtained strains is the low substrate specificity of the basic CoA-dependent alcohol/aldehyde AdhE dehydrogenase from E. coli to butyryl-CoA. It was concluded that, in order to construct an efficient 1-butanol producer based on a model strain synthesizing butyryl-CoA as a result of a reversal in fatty acid beta-oxidation enzymes, it is necessary to provide intensive formation of acetyl-CoA and enhanced activity of alternative alcohol and aldehyde dehydrogenases in the cells of a strain.
Product Description
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, BioRenewable, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG