Merck
  • Construction of the octose 8-phosphate intermediate in lincomycin A biosynthesis: characterization of the reactions catalyzed by LmbR and LmbN.
Construction of the octose 8-phosphate intermediate in lincomycin A biosynthesis: characterization of the reactions catalyzed by LmbR and LmbN.
Journal of the American Chemical Society (2012-09-20)
Eita Sasaki, Chia-I Lin, Ke-Yi Lin, Hung-Wen Liu
ABSTRACT
Lincomycin A is a potent antimicrobial agent noted for its unusual C1 methylmercapto-substituted 8-carbon sugar. Despite its long clinical history for the treatment of Gram-positive infections, the biosynthesis of the C(8)-sugar, methylthiolincosamide (MTL), is poorly understood. Here, we report our studies of the two initial enzymatic steps in the MTL biosynthetic pathway leading to the identification of D-erythro-D-gluco-octose 8-phosphate as a key intermediate. Our experiments demonstrate that this intermediate is formed via a transaldol reaction catalyzed by LmbR using D-fructose 6-phosphate or D-sedoheptulose 7-phosphate as the C(3) donor and D-ribose 5-phosphate as the C(5) acceptor. Subsequent 1,2-isomerization catalyzed by LmbN converts the resulting 2-keto C(8)-sugar (octulose 8-phosphate) to octose 8-phosphate. These results provide, for the first time, in vitro evidence for the biosynthetic origin of the C(8) backbone of MTL.
Product Description
Lincomycin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Lincomycin hydrochloride monohydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Lincomycin hydrochloride, 96.0-102.0% (HPLC)
Sigma-Aldrich
Lincomycin hydrochloride, ≥90% (TLC)
Sigma-Aldrich
Lincomycin hydrochloride, BioReagent, suitable for cell culture