Merck
  • Biological activity and safety of Tripterygium extract prepared by sodium carbonate extraction.
Biological activity and safety of Tripterygium extract prepared by sodium carbonate extraction.
Molecules (Basel, Switzerland) (2012-09-19)
Wei Fang, Fan Peng, Tao Yi, Cong Zhang, Chunxi Wan, Huibi Xu, Christopher Waikei Lam, Xiangliang Yang
ABSTRACT
The commercial preparation named “Tripterygium glycosides” prepared by column chromatography has been used for the treatment of inflammatory and autoimmune diseases with significant efficacy but concurrent toxicity. The aim of this study was to reduce the toxicity of Tripterygium extracts, using cytotoxicity and anti-inflammatory activity of the three principal active components of Tripterygium wilfordii Hook. F. (TWHF)as guiding parameters. Column chromatography was replaced by sodium carbonate extraction for removing the acidic compounds and enriching epoxyditerpenoids and alkaloids in the extract. Results showed that the therapeutic index (IC50/EC50) on murine macrophage Raw 264.7 cells and rat mesangial HBZY-1 cells of the extract prepared by sodium carbonate extraction was significantly higher than that of Tripterygium glycosides(0.8 and 5.2 vs. 0.3 and 2.6, p < 0.05), while its cytotoxicity on human liver HL7702 cells was significantly lower (14.5 ± 1.4 vs. 6.8 ± 0.9, p < 0.05). Further acute oral toxicity experiments showed that the LD50 value of this extract was 1,210 mg/kg compared to 257 mg/kg for Tripterygium glycosides. All the above results suggest that Tripterygium extract prepared by sodium carbonate extraction may represent a potentially optimal source of medicine with good therapeutic index.
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate decahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Triptolide, from Tripterygium wilfordii, ≥98% (HPLC), solid
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate decahydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate monohydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, 99.5-100.5% (ex dried subst.)
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate monohydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate monohydrate, ReagentPlus®, ≥99.5%
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
Celastrol, ≥98% (HPLC), solid
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C