Merck
  • A novel sample preparation method using rapid nonheated saponification method for the determination of cholesterol in emulsified foods.
A novel sample preparation method using rapid nonheated saponification method for the determination of cholesterol in emulsified foods.
Journal of food science (2012-09-18)
In-Seek Jeong, Byung-Man Kwak, Jang-Hyuk Ahn, Donggil Leem, Taehyung Yoon, Changyong Yoon, Jayoung Jeong, Jung-Min Park, Jin-Man Kim
ABSTRACT
In this study, nonheated saponification was employed as a novel, rapid, and easy sample preparation method for the determination of cholesterol in emulsified foods. Cholesterol content was analyzed using gas chromatography with a flame ionization detector (GC-FID). The cholesterol extraction method was optimized for maximum recovery from baby food and infant formula. Under these conditions, the optimum extraction solvent was 10 mL ethyl ether per 1 to 2 g sample, and the saponification solution was 0.2 mL KOH in methanol. The cholesterol content in the products was determined to be within the certified range of certified reference materials (CRMs), NIST SRM 1544 and SRM 1849. The results of the recovery test performed using spiked materials were in the range of 98.24% to 99.45% with an relative standard devitation (RSD) between 0.83% and 1.61%. This method could be used to reduce sample pretreatment time and is expected to provide an accurate determination of cholesterol in emulsified food matrices such as infant formula and baby food. A novel, rapid, and easy sample preparation method using nonheated saponification was developed for cholesterol detection in emulsified foods. Recovery tests of CRMs were satisfactory, and the recoveries of spiked materials were accurate and precise. This method was effective and decreased the time required for analysis by 5-fold compared to the official method.
Product Description
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder