Merck
  • Simultaneous determination of amlodipine and bisoprolol in rat plasma by a liquid chromatography/tandem mass spectrometry method and its application in pharmacokinetic study.
Simultaneous determination of amlodipine and bisoprolol in rat plasma by a liquid chromatography/tandem mass spectrometry method and its application in pharmacokinetic study.
Journal of pharmaceutical and biomedical analysis (2012-09-06)
Huichao Chang, Jinyin Li, Ji Li, Xiaoduo Guan, Fanlu Sun, Zhongzhi Qian, Kaishun Bi, Guorong Fan
ABSTRACT
A sensitive, specific liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was established for the quantitative determination of amlodipine and bisoprolol, using clenbuterol as the internal standard (IS). The analytes and IS were isolated from 100μL plasma samples by a simple liquid-liquid extraction (LLE). Reverse-phase high performance liquid chromatography (RP-HPLC) separation was accomplished on a Diamonsil C(18) column (50mm×4.6mm, 5μm) with a mobile phase composed of methanol-water-formic acid (75:25:0.01, v/v/v) at a flow rate of 0.3mL/min. The method had a chromatographic total run time of 3min. Multiple reacting monitoring (MRM) transitions of m/z [M+H](+) 409.1→237.9 (amlodipine), m/z [M+H](+) 326.2→116.0 (bisoprolol) and m/z [M+H](+) 277.0→203.0 (clenbuterol, IS) were used to quantify amlodipine, bisoprolol and IS, respectively. The method was sensitive with a lower limit of quantitation (LLOQ) of 0.2ng/mL for both amlodipine and bisoprolol, and the linear range was 0.2-50ng/mL for both amlodipine and bisoprolol (r(2)>0.9961). All the validation data, such as accuracy, precision and inter-day repeatability, were within the required limits. The method was successfully applied to pharmacokinetic studies of amlodipine and bisoprolol in Sprague-Dawley (SD) rats.
Product Description
Bisoprolol for system suitability, European Pharmacopoeia (EP) Reference Standard
Bisoprolol fumarate, European Pharmacopoeia (EP) Reference Standard
Bisoprolol for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bisoprolol hemifumarate salt, ≥98% (HPLC), solid