Merck
  • A new, direct analytical method using LC-MS/MS for fatty acid esters of 3-chloro-1,2-propanediol (3-MCPD esters) in edible oils.
A new, direct analytical method using LC-MS/MS for fatty acid esters of 3-chloro-1,2-propanediol (3-MCPD esters) in edible oils.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2012-08-23)
K Yamazaki, M Ogiso, S Isagawa, T Urushiyama, T Ukena, N Kibune
ABSTRACT
A new, direct analytical method for the determination of 3-chloro-1,2-propanediol fatty acid esters (3-MCPD esters) was developed. The targeted 3-MCPD esters included five types of monoester and 25 [corrected] types of diester. Samples (oils and fats) were dissolved in a mixture of tert-butyl methyl ether and ethyl acetate (4:1), purified using two solid-phase extraction (SPE) cartridges (C(18) and silica), then analysed by liquid chromatography-tandem mass spectrometry (LC-MS/MS). Five monoesters and five diesters with the same fatty acid group could be separated and quantified. Pairs of 3-MCPD diesters carrying the same two different fatty acid groups, but at reversed positions (sn-1 and sn-2), could not be separated and so were expressed as a sum of both compounds. The limits of quantification (LOQs) were estimated to be between 0.02 to 0.08 mg kg(-1), depending on the types of 3-MCPD ester. Repeatability expressed as relative standard deviation (RSD(r)%) varied from 5.5% to 25.5%. The new method was shown to be applicable to various commercial edible oils and showed levels of 3-MCPD esters varying from 0.58 to 25.35 mg kg(-1). The levels of mono- and diesters ranged from 0.10 to 0.69 mg kg(-1) and from 0.06 to 16 mg kg(-1), respectively.
Sigma-Aldrich
(R)-(−)-3-Chloro-1,2-propanediol, 97%, optical purity ee: 98% (GLC)
Sigma-Aldrich
(±)-3-Chloro-1,2-propanediol, 98%
Sigma-Aldrich
(S)-(+)-3-Chloro-1,2-propanediol, 97%, optical purity ee: 97% (GLC)
Supelco
(±)-3-Chloro-1,2-propanediol, analytical standard