Merck
  • Nanocarriers for dermal drug delivery: influence of preparation method, carrier type and rheological properties.
Nanocarriers for dermal drug delivery: influence of preparation method, carrier type and rheological properties.
International journal of pharmaceutics (2012-08-21)
Julia C Schwarz, Angelika Weixelbaum, Elisabeth Pagitsch, Monika Löw, Guenter P Resch, Claudia Valenta
ABSTRACT
Nanocarriers are highly interesting delivery systems for the dermal application of drugs. Based on a eudermic alkylpolyglycosid nanoemulsions, solid lipid nanoparticles (SLN) and nano-structured lipid carriers (NLC) were prepared by ultrasonic dispersion. The ultrasound preparation technique turned out to be convenient and rapid. For reasons of comparison, nanoemulsions were also prepared by high-pressure homogenisation with highly similar physicochemical properties. Cryo electron microscopy was employed to elucidate the microstructure of the ultrasound-engineered nanocarriers. Furthermore, in vitro skin experiments showed excellent skin permeation and penetration properties for flufenamic acid from all formulations. Moreover, ATR-FTIR studies revealed barrier-restorative properties for NLC and SLN. Furthermore, the rheological characteristics of all nanocarriers were determined. In order to increase the viscosity, three different polymers were employed to also prepare semi-solid NLC drug delivery systems. All of them exhibited comparable skin diffusion properties, but may offer improved dermal applicability.
Product Description
Supelco
Flufenamic acid, analytical standard, for drug analysis