Merck
  • Micelles based on methoxy poly(ethylene glycol)-cholesterol conjugate for controlled and targeted drug delivery of a poorly water soluble drug.
Micelles based on methoxy poly(ethylene glycol)-cholesterol conjugate for controlled and targeted drug delivery of a poorly water soluble drug.
Journal of biomedical nanotechnology (2012-08-15)
Junming Li, Zhiyao He, Shui Yu, Shuangzhi Li, Qing Ma, Yiyi Yu, Jialin Zhang, Rui Li, Yu Zheng, Gu He, Xiangrong Song
ABSTRACT
In this study, quercetin (QC) with cancer chemoprevention effect and anticancer potential was loaded into polymeric micelles of methoxy poly(ethylene glycol)-cholesterol conjugate (mPEG-Chol) in order to increase its water solubility. MPEG-Chol with lower critical micelle concentration (CMC) value (4.0 x 10(-7) M - 13 x 10(-7) M) was firstly synthesized involving two steps of chemical modification on cholesterol by esterification, and then QC was incorporated into mPEG-Chol micelles by self-assembly method. After the process parameters were optimized, QC-loaded micelles had higher drug loading (3.66%) and entrapment efficiency (93.51%) and nano-sized diameter (116 nm). DSC analysis demonstrated that QC had been incorporated non-covalently into the micelles and existed as an amorphous state or a solid solution in the polymeric matrix. The freeze-dried formulation with addition of 1% (w/v) mannitol as cryoprotectant was successfully developed for the long-term storage of QC-loaded micelles. Compared to free QC, QC-loaded micelles could release QC more slowly. Moreover, the release of QC from micelles was slightly faster in PBS at pH 5 than that in PBS at pH 7.4, which implied that QC-loaded micelles might be pH-sensitive and thereby selectively deliver QC to tumor tissue with unwanted side effects. Therefore, mPEG-Chol was a promising micellar vector for the controlled and targeted drug delivery of QC to tumor and QC-loaded micelles were also worth being further investigated as a potential formulation for cancer chemoprevention and treatment.
Sigma-Aldrich
Methoxypolyethylene glycol 350, average mol wt 350
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 550
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn ~2,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 750
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, BioUltra, 500
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 5,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 20,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 10,000