Merck
  • Efficacy of intraoperative, single-bolus corticosteroid administration to prevent postoperative acute respiratory failure after oesophageal cancer surgery.
Efficacy of intraoperative, single-bolus corticosteroid administration to prevent postoperative acute respiratory failure after oesophageal cancer surgery.
Interactive cardiovascular and thoracic surgery (2012-06-30)
Seong Yong Park, Hyun-Sung Lee, Hee-Jin Jang, Jungnam Joo, Jae Ill Zo
ABSTRACT
Respiratory failure from acute lung injury (ALI), acute respiratory distress syndrome (ARDS) and pneumonia are the major cause of morbidity and mortality following an oesophagectomy for oesophageal cancer. This study was performed to investigate whether an intraoperative corticosteroid can attenuate postoperative respiratory failure. Between November 2005 and December 2008, 234 consecutive patients who underwent an oesophagectomy for oesophageal cancer were reviewed. A 125-mg dose of methylprednisolone was administered after performing the anastomosis. ALI, ARDS and pneumonia occurring before postoperative day (POD) 7 were regarded as acute respiratory failure. The mean age was 64.2 ± 8.7 years. One hundred and fifty-one patients were in the control group and 83 patients in the steroid group. Patients' characteristics were comparable. The incidence of acute respiratory failure was lower in the steroid group (P = 0.037). The incidences of anastomotic leakage and wound dehiscence were not different (P = 0.57 and P = 1.0). The C-reactive protein level on POD 2 was lower in the steroid group (P < 0.005). Multivariate analysis indicates that the intraoperative steroid was a protective factor against acute respiratory failure (P = 0.046, OR = 0.206). Intraoperative corticosteroid administration was associated with a decreased risk of acute respiratory failure following an oesophagectomy. The laboratory data suggest that corticosteroids may attenuate the stress-induced inflammatory responses after surgery.
Sigma-Aldrich
6α-Methylprednisolone 21-hemisuccinate sodium salt, lyophilized powder
USP
Methylprednisolone hemisuccinate, United States Pharmacopeia (USP) Reference Standard
Methylprednisolone hydrogen succinate for performance test, European Pharmacopoeia (EP) Reference Standard
Methylprednisolone hydrogen succinate, European Pharmacopoeia (EP) Reference Standard