Merck
  • Pretreatment of sugarcane bagasse with NH4OH-H2O2 and ionic liquid for efficient hydrolysis and bioethanol production.
Pretreatment of sugarcane bagasse with NH4OH-H2O2 and ionic liquid for efficient hydrolysis and bioethanol production.
Bioresource technology (2012-06-26)
Zhisheng Zhu, Mingjun Zhu, Zhenqiang Wu
ABSTRACT
An efficient pretreatment method using NH(4)OH-H(2)O(2) and ionic liquid (IL) was developed for the recovery of cellulose from sugarcane bagasse (SCB). The regenerated SCB from the combined pretreatment exhibited significantly enhanced enzymatic digestibility with an efficiency of 91.4% after 12h of hydrolysis, which was 64% higher than the efficiency observed for the regenerated SCB after the individual NH(4)OH-H(2)O(2) pretreatment. 1-Allyl-3-methylimidazolium chloride ([Amim]Cl) dissolved the cellulose from the NH(4)OH-H(2)O(2)-pretreated SCB, and the crystallinity index (CrI) detected by X-ray diffraction (XRD) was reduced by 42%. The recycled and fresh [Amim]Cl demonstrated the same performance on the pretreatment of SCB for the enhancement of enzymatic digestibility. The regenerated SCB was subsequently used in simultaneous saccharification and co-fermentation (SScF) for bioethanol production by cellulase and yeast. The pretreatment did not have a negative effect on bioethanol fermentation, and an ethanol yield of 0.42 g/g was achieved with a corresponding fermentation efficiency of 94.5%.
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
Sigma-Aldrich
Ammonium hydroxide solution, puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a. plus, ≥25% NH3 in H2O
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis