Merck
  • Improved performance of diatomite-based dental nanocomposite ceramics using layer-by-layer assembly.
Improved performance of diatomite-based dental nanocomposite ceramics using layer-by-layer assembly.
International journal of nanomedicine (2012-05-24)
Xiaoli Lu, Yang Xia, Mei Liu, Yunzhu Qian, Xuefeng Zhou, Ning Gu, Feimin Zhang
ABSTRACT
To fabricate high-strength diatomite-based ceramics for dental applications, the layer-by-layer technique was used to coat diatomite particles with cationic [poly(allylamine hydrochloride)] and anionic [poly(sodium 4-styrenesulfonate)] polymers to improve the dispersion and adsorption of positively charged nano-ZrO(2) (zirconia) as a reinforcing agent. The modified diatomite particles had reduced particle size, narrower size distribution, and were well dispersed, with good adsorption of nano-ZrO(2). To determine the optimum addition levels for nano-ZrO(2), ceramics containing 0, 20, 25, 30, and 35 wt% nano-ZrO(2) were sintered and characterized by the three-point bending test and microhardness test. In addition to scanning electron microscopy, propagation phase-contrast synchrotron X-ray microtomography was used to examine the internal structure of the ceramics. The addition of 30 wt% nano-ZrO(2) resulted in the highest flexural strength and fracture toughness with reduced porosity. Shear bond strength between the core and veneer of our diatomite ceramics and the most widely used dental ceramics were compared; the shear bond strength value for the diatomite-based ceramics was found to be significantly higher than for other groups (P < 0.05). Our results show that diatomite-based nanocomposite ceramics are good potential candidates for ceramic-based dental materials.
Product Description
Supelco
Celatom®, acid-washed, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
Celpure® P65, meets USP/NF testing specifications
Sigma-Aldrich
Celite® 503, filter aid, treated with sodium carbonate, calcined
Supelco
Celite® 545 AW, reagent grade
Sigma-Aldrich
Hyflo® Super Cel®, filter aid, flux calcined, treated with sodium carbonate
Sigma-Aldrich
Diatomaceous earth non-washed, powder
Sigma-Aldrich
Celite® 545 AW, filter aid, acid washed, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Celite® 535, filter aid, flux calcined
Sigma-Aldrich
Filter agent, Celite® 545
Sigma-Aldrich
Diatomaceous earth
Sigma-Aldrich
Celpure® P1000, meets USP/NF testing specifications
Sigma-Aldrich
Celite® S, filter aid, dried, untreated
Sigma-Aldrich
Celite® 545, filter aid, treated with sodium carbonate, flux calcined