Merck
  • Curcumin loaded mixed micelles composed of Pluronic P123 and F68: preparation, optimization and in vitro characterization.
Curcumin loaded mixed micelles composed of Pluronic P123 and F68: preparation, optimization and in vitro characterization.
Colloids and surfaces. B, Biointerfaces (2012-05-23)
Liyan Zhao, Jianchao Du, Yuwei Duan, Ya'ni Zang, Huaisong Zhang, Chunfen Yang, Fengliang Cao, Guangxi Zhai
ABSTRACT
In this study, curcumin (Cur) loaded mixed micelles (Cur-PF), composed of Pluronic P123 (P123) and Pluronic F68 (F68), was prepared using the thin-film hydration method and evaluated in vitro. The preparation process was optimized with a central composite design (CCD). The average size of the mixed micelles was 68.2 nm, and the encapsulating efficiency for Cur was 86.93%, and 6.996% for drug-loading. Compared with the Cur propylene glycol solution, the in vitro release of Cur from Cur-PF presented the sustained-release property. The in vitro cytotoxicity assay showed that the IC(50) values on MCF-7 cells for Cur-PF and free Cur in DMSO solution were 5.04 μg/mL and 8.35 μg/mL, while 2.52 μg/mL and 8.27 μg/mL on MCF-7/ADR cells. It could be concluded from the results that P123/F68 mixed micelles might serve as a potential nanocarrier to improve the solubility and biological activity of Cur.
Sigma-Aldrich
Poloxamer 188, micronized
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,000
Sigma-Aldrich
Poloxamer 188 solution, 10%, sterile-filtered, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Poloxamer 407, micronized
Sigma-Aldrich
Poloxamer 188
Sigma-Aldrich
Poloxamer 188, solid
Sigma-Aldrich
Poloxamer 407, purified, non-ionic
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Synperonic® PE P105, surfactant
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,100
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,900
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,900
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~4,400
Sigma-Aldrich
Synperonic® F 108, surfactant, non-ionic
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~3,300
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~14,600
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~8,400
Sigma-Aldrich
Poloxamer 407, oxyethylene 71.5-74.9 %
Sigma-Aldrich
Synperonic® PE/P84
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~5,800