- Living Fasciola hepatica in biliary tree: a case report.
Living Fasciola hepatica in biliary tree: a case report.
Annals of hepatology (2012-04-07)
Keyvan Aminian, Kambiz Akhavan Rezayat, Afshin Shafaghi, Roozbeh Tanhaeevash
PMID22481460
ABSTRACT
Here we report a rare case of living Fasciola hepatica in biliary tract. The patient was in acute phase of infection and treated successfully with 10 mg/kg oral triclabendazole after the fluke was extracted using endoscopic retrograde cholangiopancreatography (ERCP).