Merck
[Test and analysis of acrylic acid ester in industry pipelines].
Huan jing ke xue= Huanjing kexue (2012-04-04)
Bin Wu, Hong-Yan Zhang, Lin-Guang Lu
ABSTRACT
A new quality and quantity analyzing method for waste gas from chemical industry, especially for acrylic acid ester using air-sampling combined with GC/MS and honeywell air purifier system was firstly advanced in this paper. According to the experiments, the maximum disposal capability of the honeywell air purifier system for acrylic acid ester gas was calculated as about 1 kg x h(-1) and the feasibility and validity of the method was also proved by parallel tests.
Product Description
Supelco
Butyl acrylate, analytical standard
Sigma-Aldrich
Butyl acrylate, ≥99%, contains 10-60 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Ethyl acrylate, contains 10-20 ppm MEHQ as inhibitor, 99%
Supelco
Ethyl acrylate, analytical standard
Sigma-Aldrich
Ethyl acrylate, ≥99.5%, stabilized