Merck
  • Salbutamol but not ipratropium abolishes leukotriene D4-induced gas exchange abnormalities in asthma.
Salbutamol but not ipratropium abolishes leukotriene D4-induced gas exchange abnormalities in asthma.
European journal of clinical pharmacology (2012-03-30)
Barbro Dahlén, Federico P Gómez, Alejandro Casas, Peter H Howarth, Sven-Erik Dahlén, Robert Rodriguez-Roisin
ABSTRACT
Leukotriene D(4) (LTD(4)) is a central mediator in asthma inducing bronchoconstriction and profound disturbances in pulmonary gas exchange in asthmatic subjects. The aim of the study was to compare, for the first time, the influence of the bronchodilators salbutamol (400 μg) and ipratropium (80 μg) on lung function changes induced by inhaled LTD(4). Treatments were evaluated in a randomized, three-period, double-blind, placebo-controlled, cross-over study where spirometric and pulmonary gas exchange indices were followed in 12 subjects with mild asthma before and after LTD(4) challenge. Compared with placebo, salbutamol provided significant protection against the fall in FEV(1) (forced expiratory volume in 1 s) after LTD(4) challenge. Salbutamol also abolished the LTD(4)-induced gas exchange disturbances [decreased arterial oxygen tension (PaO(2)) and increased alveolar-arterial oxygen tension difference (AaPO(2))]. Ipratropium provided significant but less marked attenuation of the changes in FEV(1) and arterial oxygenation induced by LTD(4). Despite the equal bronchodilatory effects of salbutamol and ipratropium before the challenge with LTD(4), salbutamol was superior to ipratropium in preventing spirometric and gas exchange abnormalities. This result indicates a broader action of salbutamol on several of the disturbances that contribute to airway obstruction including, for example, exudation of plasma in the airway mucosa. The clinical implication of this new finding is that in this model of acute asthmatic airway obstruction, salbutamol was more effective than ipratropium.
Sigma-Aldrich
Leukotriene D4, ~50 μg/mL (in methanol/ammonium acetate buffer, 70:30, pH 5.6), ≥97%