Merck
  • Non-covalent versus covalent donor-acceptor systems based on near-infrared absorbing azulenocyanines and C60 fullerene derivatives.
Non-covalent versus covalent donor-acceptor systems based on near-infrared absorbing azulenocyanines and C60 fullerene derivatives.
Chemical communications (Cambridge, England) (2012-03-22)
Mine Ince, Anita Hausmann, M Victoria Martínez-Díaz, Dirk M Guldi, Tomás Torres
ABSTRACT
A novel supramolecular electron donor-acceptor hybrid (2·1) and an electron donor-acceptor conjugate (3), both exhibiting a remarkably shifted Q band in the NIR region of the solar spectrum, were prepared. Irradiation of the supramolecular ensemble 2·1 within the visible range leads to a nanosecond lived radical-ion pair state Zn-azulenocyanine˙(+)-C(60)˙(-).
Sigma-Aldrich
Azulene, 99%