Merck
  • A new method for removal of hydrogen peroxide interference in the analysis of chemical oxygen demand.
A new method for removal of hydrogen peroxide interference in the analysis of chemical oxygen demand.
Environmental science & technology (2012-02-01)
Tingting Wu, James D Englehardt
ABSTRACT
Many advanced oxidation processes involve addition of hydrogen peroxide (H(2)O(2)) with the aim of generating hydroxyl radicals to oxidize organic contaminants in water. However, chemical oxygen demand, a common measure of gross residual organic contamination, is subject to interference from residual H(2)O(2) in the treated water. A new method, involving catalytic decomposition of H(2)O(2) with addition of heat and sodium carbonate (Na(2)CO(3)), is proposed in this work to address this problem. The method is demonstrated experimentally, and modeled kinetically. Results for 5 mM H(2)O(2) in deionized (DI) water included reduction to below the COD detection limit after 60 min heating (90(◦)C) with addition of 20 g/L Na(2)CO(3) concentrated solution, whereas 900 min were required in treated municipal wastewater. An approximate second order rate constant of 11.331 M(-1)·min(-1) at Na(2)CO(3) dosage of 20 g/L was found for the tested wastewater. However, kinetic modeling indicated a two-step reaction mechanism, with formation of peroxocarbonate (CO(4)(2-)) and ultimate decomposition to H(2)O and O(2) in pure H(2)O(2) solution. A similar mechanism is apparent in wastewater at high catalyst concentrations, whereas at low Na(2)CO(3) addition rates, the catalytic effects of other constituents appear important.
Sigma-Aldrich
Sodium carbonate monohydrate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate monohydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, 99.5-100.5% (ex dried subst.)
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate decahydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium carbonate monohydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate decahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C