Merck
Yb3+ sensitized Tm3+ upconversion in tellurite lead oxide glass.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-01-24)
Deepak Kumar Mohanty, Vineet Kumar Rai, Y Dwivedi
ABSTRACT
Triply ionized thulium/thulium--ytterbium doped/codoped TeO2-Pb3O4 (TPO) glasses have been fabricated by classical quenching method. The upconversion emission spectra in the Tm3+/Tm3+-Yb3+ doped/codoped glasses upon excitation with a diode laser lasing at ∼980 nm has been studied. Effect of the addition of the Yb3+ on the upconversion emission intensity in the visible and near infrared regions of the Tm3+ doped in TPO glass has been studied and the processes involved explored.
Sigma-Aldrich
Lead(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Lead(II) oxide, ACS reagent, ≥99.0%
Sigma-Aldrich
Lead(II) oxide, puriss. p.a., ≥99.0% (KT), yellow
Sigma-Aldrich
Lead(II) oxide, powder, <10 μm, ReagentPlus®, ≥99.9% trace metals basis