Merck
  • Interleukin-13 enhanced Ca2+ oscillations in airway smooth muscle cells.
Interleukin-13 enhanced Ca2+ oscillations in airway smooth muscle cells.
Cytokine (2011-11-15)
Hisako Matsumoto, Yutaka Hirata, Kojiro Otsuka, Toshiyuki Iwata, Aya Inazumi, Akio Niimi, Isao Ito, Emiko Ogawa, Shigeo Muro, Hiroaki Sakai, Kazuo Chin, Yoshitaka Oku, Michiaki Mishima
ABSTRACT
Physiological mechanisms associated with interleukin-13 (IL-13), a key cytokine in asthma, in intracellular Ca(2+) signaling in airway smooth muscle cells (ASMCs) remain unclear. The aim of this study was to assess effects of IL-13 on Ca(2+) oscillations in response to leukotriene D4 (LTD4) in human cultured ASMCs. LTD4-induced Ca(2+) oscillations in ASMCs pretreated with IL-13 were imaged by confocal microscopy. mRNA expressions of cysteinyl leukotriene 1 receptors (CysLT1R), CD38, involved with the ryanodine receptors (RyR) system, and transient receptor potential canonical (TRPC), involved with store-operated Ca(2+) entry (SOCE), were determined by real-time PCR. In IL-13-pretreated ASMCs, frequency of LTD4-induced Ca(2+) oscillations and number of oscillating cells were significantly increased compared with untreated ASMCs. Both xestospongin C, a specific inhibitor of inositol 1,4,5-triphosphate receptors (IP(3)R), and ryanodine or ruthenium red, inhibitors of RyR, partially blocked LTD4-induced Ca(2+) oscillations. Ca(2+) oscillations were almost completely inhibited by 50 μM of 2-aminoethoxydiphenyl borate (2-APB), which dominantly blocks SOCE but not IP(3)R at this concentration. Pretreatment with IL-13 increased the mRNA expressions of CysLT1R and CD38, but not of TRPC1 and TRPC3. We conclude that IL-13 enhances frequency of LTD4-induced Ca(2+) oscillations in human ASMCs, which may be cooperatively modulated by IP(3)R, RyR systems and possibly by SOCE.
Sigma-Aldrich
Leukotriene D4, ~50 μg/mL (in methanol/ammonium acetate buffer, 70:30, pH 5.6), ≥97%