Merck
  • Evaluation of a methacrylate-bonded cyclodextrins as a monolithic chiral stationary phase for capillary electrochromatography (CEC)-UV and CEC coupled to mass spectrometry.
Evaluation of a methacrylate-bonded cyclodextrins as a monolithic chiral stationary phase for capillary electrochromatography (CEC)-UV and CEC coupled to mass spectrometry.
Electrophoresis (2011-10-11)
Congying Gu, Shahab A Shamsi
ABSTRACT
Glycidyl methacrylate-bonded β-cyclodextrin (GMA-β-CD) is synthesized as a new chiral monomer by direct chemical bonding with GMA using a fast and simple alternative procedure. Next, rigid and homogenous monolithic columns were prepared by polymerization of GMA-β-CD monomer with ethylene dimethacrylate (EDMA), in the presence of commonly used porogens and a charged achiral monomer to form a versatile chiral monolith. This is the first report in which a preparation procedure for a methacrylate-bonded CD is introduced for chiral separations in CEC. The degree of substitution of GMA-β-CD monomer and mobile-phase parameters were optimized to achieve the highest enantioselectivity and plate number. To evaluate the GMA-β-CD monolithic column, different classes of chiral compounds were screened. Under the optimized β-CD monolith phase and the optimum mobile-phase conditions, 30 neutral and basic chiral compounds and two acidic compounds could be separated. The high chemical and mechanical stability, homogenous microflow and no loss of material at the interface allows for the first time the feasibility of applying this polymer-based monolithic column for CEC coupled to ESI-MS. Compared with CEC-UV, CEC-ESI-MS showed higher sensitivity and lower resolution. However, resolution greater than 1.0 can still be obtained for majority of the select tested compound in CEC-ESI-MS with at least three out of seven compound providing Rs≥1.5. The results reinforce the potential of GMA-β-CD monolithic columns for chiral separations with high sensitivity in CEC-ESI-MS. Finally, using hexobarbital as the model chiral analyte, the monolithic column demonstrated excellent stability and reproducibility of retention time and enantioselectivity.
Sigma-Aldrich
Glycidyl methacrylate, ≥97.0% (GC)
Sigma-Aldrich
Glycidyl methacrylate, 97%, contains 100 ppm monomethyl ether hydroquinone as inhibitor