Merck
  • Knock-out of the potassium channel TASK-1 leads to a prolonged QT interval and a disturbed QRS complex.
Knock-out of the potassium channel TASK-1 leads to a prolonged QT interval and a disturbed QRS complex.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2011-08-26)
Niels Decher, Konstantin Wemhöner, Susanne Rinné, Michael F Netter, Marylou Zuzarte, Maria I Aller, Susann G Kaufmann, Xian Tao Li, Sven G Meuth, Jürgen Daut, Frank B Sachse, Sebastian K G Maier
ABSTRACT
The aim of the study was to characterize the whole cell current of the two-pore domain potassium channel TASK-1 (K2P3) in mouse ventricular cardiomyocytes (I(TASK-1)) and to analyze the cardiac phenotype of the TASK-1(-/-) mice. We have quantified the ventricular I(TASK-1) current using the blocker A293 and TASK-1(-/-) mice. Surface electrocardiogram recordings of TASK-1(-/-) mice showed a prolonged QTc interval and a broadened QRS complex. The differences in electrocardiograms between wild type and TASK-1(-/-) mice disappeared during sympathetic stimulation of the animals. Quantitative RT-PCR, patch clamp recordings and measurements of hemodynamic performance of TASK-1(-/-) mice revealed no major compensatory changes in ion channel transcription. Action potential recordings of TASK-1(-/-) mouse cardiomyocytes indicated that I(TASK-1) modulates action potential duration. Our in vivo electrophysiological studies showed that isoflurane, which activates TASK-1, slowed heart rate and atrioventricular conduction of wild-type but not of TASK-1(-/-) mice. The results of an invasive electrophysiological catheter protocol in combination with the observed QRS time prolongation in the surface electrocardiogram point towards a regulatory role of TASK-1 in the cardiac conduction system.
Sigma-Aldrich
Methoxamine hydrochloride