Merck
  • Pharmacokinetics of caffeine in plasma and saliva, and the influence of caffeine abstinence on CYP1A2 metrics.
Pharmacokinetics of caffeine in plasma and saliva, and the influence of caffeine abstinence on CYP1A2 metrics.
The Journal of pharmacy and pharmacology (2011-08-11)
Vidya Perera, Annette S Gross, Hongmei Xu, Andrew J McLachlan
ABSTRACT
To investigate the utility of metrics of CYP1A2 activity using caffeine as a probe, and saliva and plasma sampling with or without a 24-h caffeine abstinence. This was a cross-over pharmacokinetic study in 30 healthy male subjects who received a single oral 100mg caffeine dose after 24-h caffeine abstinence or after maintaining their regular caffeine intake (no caffeine abstinence). Serial blood and saliva samples were collected simultaneously over 24h. Caffeine and paraxanthine concentrations were measured using a validated HPLC assay. There was a strong correlation between the paraxanthine/caffeine AUC(0-24) ratio (reference metric) and the paraxanthine/caffeine concentration (C(t) ) ratio at 4h (C(4) ) in both saliva and plasma (r≥0.75). The paraxanthine/caffeine AUC(0-24) ratio in plasma and saliva did not differ between the 24-h caffeine abstinence and the no abstinence period (P>0.05). The optimal paraxanthine/caffeine C(t) that correlated with the plasma paraxanthine/caffeine AUC(0-24) ratio in the 24-h abstinence period was 2 and 4h (r=0.88) in plasma, and 4 and 6h in saliva (r=0.70), while it was the saliva 4h time-point in the no abstinence period (r=0.78). The saliva paraxanthine/caffeine concentration ratio at 4h was a suitable metric to assess CYP1A2 activity after oral administration of caffeine without the need for 24-h caffeine abstinence.
Product Description
Supelco
Paraxanthine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
1,7-Dimethylxanthine, ~98%, solid