Merck
  • Development of a new single-bottle multi-purpose primer for bonding to dental porcelain, alumina, zirconia, and dental gold alloy.
Development of a new single-bottle multi-purpose primer for bonding to dental porcelain, alumina, zirconia, and dental gold alloy.
Dental materials journal (2011-07-23)
Kunio Ikemura, Hisaki Tanaka, Toshihide Fujii, Mikito Deguchi, Takeshi Endo, Yoshinori Kadoma
ABSTRACT
This study investigated the bonding efficacy of a combined primer application which comprised a silane coupling agent, an acidic adhesive monomer, and a dithiooctanoate monomer, as well as the influence of shelf life on bonding. Five experimental primers (coded as Si-P-SS-1 to Si-P-SS-4, and Si-SS as the control) were prepared using 20.0-40.0 wt% 3-methacryloyloxypropyltriethoxysilane (3-MPTES), 0-7.44 wt% 6-methacryloyloxyhexyl phosphonoacetate (6-MHPA), and 0.50 wt% 10-methacryloyloxydecyl 6,8-dithiooctanoate (10-MDDT). After 24-hour storage at 23°C (Initial) and 2-month storage at 50°C (Aged), tensile bond strengths (TBSs) of a resin cement (ResiCem, Shofu Inc., Kyoto, Japan) to primer-treated porcelain, alumina, zirconia, and Au alloy were measured. With the Initial and Aged primers of Si-P-SS-1 to Si-P-SS-3, there were no statistically significant differences in the mean TBSs (MPa) [porcelain: 21.7-29.2; alumina: 21.4-25.3; zirconia: 20.3-24.5; and Au alloy: 23.4-27.6] among these three primers (p>0.05), but they were significantly higher than that of the control primer (p<0.05). The experimental primers Si-P-SS-1 to Si-P-SS-3 demonstrated good potential as multi-purpose primers: they had good shelf lives as single-bottle primer systems and were thus able to exhibit good bond strength to all the adherends tested after 2-month storage under accelerated aging conditions.
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, 98%
Sigma-Aldrich
Phosphonoacetic acid, 98%
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, ≥97%