Merck
  • New salicylamide and sulfonamide derivatives of quinoxaline 1,4-di-N-oxide with antileishmanial and antimalarial activities.
New salicylamide and sulfonamide derivatives of quinoxaline 1,4-di-N-oxide with antileishmanial and antimalarial activities.
Bioorganic & medicinal chemistry letters (2011-07-05)
Carlos Barea, Adriana Pabón, Denis Castillo, Mirko Zimic, Miguel Quiliano, Silvia Galiano, Silvia Pérez-Silanes, Antonio Monge, Eric Deharo, Ignacio Aldana
ABSTRACT
Continuing with our efforts to identify new active compounds against malaria and leishmaniasis, 14 new 3-amino-1,4-di-N-oxide quinoxaline-2-carbonitrile derivatives were synthesized and evaluated for their in vitro antimalarial and antileishmanial activity against Plasmodium falciparum Colombian FCR-3 strain and Leishmania amazonensis strain MHOM/BR/76/LTB-012A. Further computational studies were carried out in order to analyze graphic SAR and ADME properties. The results obtained indicate that compounds with one halogenous group substituted in position 6 and 7 provide an efficient approach for further development of antimalarial and antileishmanial agents. In addition, interesting ADME properties were found.
Sigma-Aldrich
O-Acetylsalicyloyl chloride, 95%
Sigma-Aldrich
Salicylamide, puriss., ≥99.0% (T)
Sigma-Aldrich
Salicylamide, 99%