Merck
Permanent press allergy in an active duty U.S. Army soldier.
Military medicine (2011-06-04)
Shelley L Aldrich, Michael R Murchland, J Scott Henning
ABSTRACT
Ethylene urea/melamine formaldehyde resin (permanent press) is a common fabric finishing agent added to Army Combat Uniforms for a wrinkle-free appearance and to strengthen the fabric. We describe the case of an active duty U.S. Army soldier with a diffuse eczematous dermatitis in whom patch testing was used to identify an allergy to permanent press, a ubiquitous fabric finishing agent in the Army combat uniform. To our knowledge, this is the first case report of a soldier with an allergic contact dermatitis to ethylene urea/melamine formaldehyde resin. This case highlights the importance of considering the diagnosis of allergic contact dermatitis in patients with a recurrent eczematous dermatitis that does not respond appropriately to therapy and the unique occupational impact of diagnosing an Army soldier with permanent press allergy.
Sigma-Aldrich
2-Imidazolidone, 96%