Merck
  • Inhibition of cell growth and induction of apoptosis in non-small cell lung cancer cells by delta-tocotrienol is associated with notch-1 down-regulation.
Inhibition of cell growth and induction of apoptosis in non-small cell lung cancer cells by delta-tocotrienol is associated with notch-1 down-regulation.
Journal of cellular biochemistry (2011-05-21)
Xiangming Ji, Zhiwei Wang, Andreea Geamanu, Fazlul H Sarkar, Smiti V Gupta
ABSTRACT
Lung cancer is the leading cause of death among all cancers. Non-small cell lung cancer accounts for 80% of lung cancer with a 5-year survival rate of 16%. Notch pathway, especially Notch-1 is up-regulated in a subgroup of non-small cell lung cancer patients. Since Notch-1 signaling plays an important role in cell proliferation, differentiation, and apoptosis, down-regulation of Notch-1 may exert anti-tumor effects. The objective of this study was to investigate whether delta-tocotrienol, a naturally occurring isoform of Vitamin E, inhibits non-small cell lung cancer cell growth via Notch signaling. Treatment with delta-tocotrienol resulted in a dose and time dependent inhibition of cell growth, cell migration, tumor cell invasiveness, and induction of apoptosis. Real-time RT-PCR and western blot analysis showed that antitumor activity by delta-tocotrienol was associated with a decrease in Notch-1, Hes-1, Survivin, MMP-9, VEGF, and Bcl-XL expression. In addition, there was a decrease in NF-κB-DNA binding activity. These results suggest that down-regulation of Notch-1, via inhibition of NF-κB signaling pathways by delta-tocotrienol, could provide a potential novel approach for prevention of tumor progression in non-small cell lung cancer.
Sigma-Aldrich
D-δ-Tocotrienol, from rice, ≥65% (GC)
Supelco
δ-Tocotrienol, analytical standard