- Synthesis of SnS nanoparticles by SILAR method for quantum dot-sensitized solar cells.
Synthesis of SnS nanoparticles by SILAR method for quantum dot-sensitized solar cells.
SnS-sensitized TiO2 electrodes were applied in quantum dot-sensitized solar cells (QDSSCs) which are environmentally more favorable than conventional Cd or Pb-chalcogenide-sensitized electrodes. SnS nanoparticles were well-distributed over the surface of TiO2 nanoparticles by the successive ionic layer adsorption and reaction (SILAR) method. Deposited SnS nanoparticles had diameter about 3 nm. Under AM1.5 irradiation with 100 mW/cm2 light intensity (at 1 sun), the energy conversion efficiency of obtained cells reached a value of 0.21% (0.25 cm2) at SILAR coating cycles of 5. In addition, the photovoltaic performance was improved by additional ZnS coating on the surface of SnS-sensitized TiO2 electrodes.