Merck
  • Review of issues concerning the use of reproductive inhibitors, with particular emphasis on resolving human-wildlife conflicts in North America.
Review of issues concerning the use of reproductive inhibitors, with particular emphasis on resolving human-wildlife conflicts in North America.
Integrative zoology (2011-03-12)
Kathleen A Fagerstone, Lowell A Miller, Gary Killian, Christi A Yoder
ABSTRACT
This manuscript provides an overview of past wildlife contraception efforts and discusses the current state of research. Two fertility control agents, an avian reproductive inhibitor containing the active ingredient nicarbazin and an immunocontraceptive vaccine, have received regulatory approval with the Environmental Protection Agency and are commercially available in the USA. OvoControl G Contraceptive Bait for Canada Geese and Ovo Control for pigeons are delivered as oral baits. An injectable immunocontraceptive vaccine (GonaCon Immunocontraceptive Vaccine) was registered with the Environmental Protection Agency for use in female white-tailed deer in September 2009. An injectable product (GonaCon Immunocontraceptive Vaccine) is registered for use in female white-tailed deer. Both products are labeled for use in urban/suburban areas where these species are overabundant. Several other compounds are currently being tested for use in wildlife in the USA, Europe, Australia and New Zealand that could have promise in the future. The development and use of reproductive inhibitors for resolving human-wildlife conflicts will depend on a number of factors, including meeting the requirements of regulatory agencies for use in the environment and on the biological and economical feasibility of their use. Use will also be dependent on health and safety issues and on public acceptance of the techniques.
Product Description
Supelco
Nicarbazin, VETRANAL®, analytical standard