Merck
  • Lewis acid/base catalyzed [2+2]-cycloaddition of sulfenes and aldehydes: a versatile entry to chiral sulfonyl and sulfinyl derivatives.
Lewis acid/base catalyzed [2+2]-cycloaddition of sulfenes and aldehydes: a versatile entry to chiral sulfonyl and sulfinyl derivatives.
Chemistry (Weinheim an der Bergstrasse, Germany) (2011-03-03)
Florian M Koch, René Peters
ABSTRACT
The first catalytic asymmetric synthesis of β-sultones is reported. This development has enabled a rapid access to a number of highly enantioenriched biologically interesting sulfonyl and sulfinyl compound classes, which makes use of the inherent ring strain of the four-membered heterocycles. The products possess either two vicinal stereocenters, such as in β-hydroxy-sulfonamides, -sulfonates, -sulfones, -sulfonic acids, -sulfinic acids, γ-sultines, and γ-sultones or a single stereocenter, such as in α-branched alkyl or allyl sulfonic acids. This work also represents the first application of sulfene intermediates in asymmetric catalysis. The reactivity of a sulfene normally acting as an electrophile could be reverted by the formation of a nucleophilic zwitterionic sulfene-amine adduct. To achieve a combination of high enantioselectivity and reactivity, cooperative catalytic action of a chiral nucleophilic tertiary amine (the cinchona alkaloid derivative diydroquinine 2,5-diphenyl-4,6-pyrimidinediyl diether ((DHQ)(2)PYR)) and Bi(OTf)(3) or In(OTf)(3) was of primary importance.
Sigma-Aldrich
Sulfamerazine, ReagentPlus®, ≥99.0%
Supelco
Sulfamerazine, VETRANAL®, analytical standard