Merck
  • Measurements of plasma methoxytyramine, normetanephrine, and metanephrine as discriminators of different hereditary forms of pheochromocytoma.
Measurements of plasma methoxytyramine, normetanephrine, and metanephrine as discriminators of different hereditary forms of pheochromocytoma.
Clinical chemistry (2011-01-26)
Graeme Eisenhofer, Jacques W M Lenders, Henri Timmers, Massimo Mannelli, Stefan K Grebe, Lorenz C Hofbauer, Stefan R Bornstein, Oliver Tiebel, Karen Adams, Gennady Bratslavsky, W Marston Linehan, Karel Pacak
ABSTRACT
Pheochromocytomas are rare catecholamine-producing tumors derived in more than 30% of cases from mutations in 9 tumor-susceptibility genes identified to date, including von Hippel-Lindau tumor suppressor (VHL); succinate dehydrogenase complex, subunit B, iron sulfur (Ip) (SDHB); and succinate dehydrogenase complex, subunit D, integral membrane protein (SDHD). Testing of multiple genes is often undertaken at considerable expense before a mutation is detected. This study assessed whether measurements of plasma metanephrine, normetanephrine, and methoxytyramine, the O-methylated metabolites of catecholamines, might help to distinguish different hereditary forms of the tumor. Plasma concentrations of O-methylated metabolites were measured by liquid chromatography with electrochemical detection in 173 patients with pheochromocytoma, including 38 with multiple endocrine neoplasia type 2 (MEN 2), 10 with neurofibromatosis type 1 (NF1), 66 with von Hippel-Lindau (VHL) syndrome, and 59 with mutations of SDHB or SDHD. In contrast to patients with VHL, SDHB, and SDHD mutations, all patients with MEN 2 and NF1 presented with tumors characterized by increased plasma concentrations of metanephrine (indicating epinephrine production). VHL patients usually showed solitary increases in normetanephrine (indicating norepinephrine production), whereas additional or solitary increases in methoxytyramine (indicating dopamine production) characterized 70% of patients with SDHB and SDHD mutations. Patients with NF1 and MEN 2 could be discriminated from those with VHL, SDHB, and SDHD gene mutations in 99% of cases by the combination of normetanephrine and metanephrine. Measurements of plasma methoxytyramine discriminated patients with SDHB and SDHD mutations from those with VHL mutations in an additional 78% of cases. The distinct patterns of plasma catecholamine O-methylated metabolites in patients with hereditary pheochromocytoma provide an easily used tool to guide cost-effective genotyping of underlying disease-causing mutations.
Sigma-Aldrich
3-Methoxytyramine hydrochloride, 99% (AT)
Sigma-Aldrich
3-Methoxytyramine hydrochloride, ≥95.5%, crystalline