Merck
  • Sage components enhance cell death through nuclear factor kappa-B signaling.
Sage components enhance cell death through nuclear factor kappa-B signaling.
Frontiers in bioscience (Elite edition) (2011-01-05)
Sally Joseph Deeb, Chirine Omar El-Baba, Saadia Bashir Hassan, Rolf Lennart Larsson, Hala Uthman Gali-Muhtasib
ABSTRACT
The sage components linalyl acetate (Ly) and alpha-terpineol (Te) exhibit synergistic anti-proliferative effects. We investigated the effects of Ly and Te on NF-kappaB signaling in HCT-116 colon cancer cells. Ly and Te combinations dose-dependently reduced HCT-116 viability at non-cytotoxic concentrations. Combination treatment induced 30%-60% increase in PreG1 through induction of apoptosis and necrosis. DNA binding assays revealed that combination treatment suppressed both basal and TNF-alpha-induced NF-kappaB activation. This suppression correlated with the inhibition of p65 nuclear translocation and IkappaB-alpha degradation. The lack of change in IKK expression levels or inhibition in IkappaB-alpha phosphorylation suggest the involvement of an IKK-independent mechanism. Ly and Te combination was found to downregulate the expression of NF-kappaB-regulated antiapoptotic and proliferative gene products. Separate treatments and drug combinations significantly decreased DNA binding activity of NF-kappaB which led to the potentiation of cell death induced by the colon cancer drugs oxaliplatin and 5-FU. These results indicate that Ly and Te anticancer activities are partly mediated through the suppression of NF-kappaB activation, suggesting their use in combination with chemotherapeutic agents to induce apoptosis.
Sigma-Aldrich
Linalyl acetate, natural, ≥96%, FG
Sigma-Aldrich
Linalyl acetate, ≥97%, FCC, FG
Supelco
(+)-α-Terpineol, analytical standard
Sigma-Aldrich
Terpineol, mixture of isomers, 96%, FG
Sigma-Aldrich
(−)−α-Terpineol, natural, ≥96%, FCC, FG
Sigma-Aldrich
α-Terpineol, 90%, technical grade
Sigma-Aldrich
3,7-Dimethyl-1,6-octadien-3-yl acetate, 97%