Merck
Total synthesis of bryostatin 1.
Journal of the American Chemical Society (2010-12-24)
Gary E Keck, Yam B Poudel, Thomas J Cummins, Arnab Rudra, Jonathan A Covel
ABSTRACT
Bryostatin 1 is a marine natural product that is a very promising lead compound because of the potent biological activity it displays against a variety of human disease states. We describe herein the first total synthesis of this agent. The synthetic route adopted is a highly convergent one in which the preformed, heavily functionalized pyran rings A and C are united by "pyran annulation", the TMSOTf-promoted reaction between a hydroxyallylsilane appended to the A-ring fragment and an aldehyde contained in the C-ring fragment, with concomitant formation of the B ring. Further elaborations of the resulting very highly functionalized intermediate include macrolactonization and selective cleavage of just one of five ester linkages present.
Sigma-Aldrich
Bryostatin 1, ≥99%, solid