Merck
  • Bromodimethylsulfonium bromide (BDMS) mediated dithioacetalization of carbohydrates under solvent-free conditions.
Bromodimethylsulfonium bromide (BDMS) mediated dithioacetalization of carbohydrates under solvent-free conditions.
Carbohydrate research (2010-09-08)
Abu T Khan, Md Musawwer Khan
ABSTRACT
A variety of diethyl dithioacetals of sugars can be prepared in very good yields by the reaction of various monosaccharides with ethanethiol in the presence of 3 mol% bromodimethylsulfonium bromide (BDMS) at 0-5°C. Similarly, dipropyl dithioacetal derivatives can also be obtained in good yields using propanethiol under identical reaction conditions. These dithioacetal derivatives were characterized by per-O-acetylation using silica gel-supported perchloric acid. The significant features of the present protocol are good-to-excellent yields, mild, clean, and solvent-free reaction conditions. This method is extremely suitable for the large-scale preparation of dithioacetal derivatives of various sugars.
Sigma-Aldrich
Propyl mercaptan, ≥97%
Sigma-Aldrich
Ethanethiol, 97%
Sigma-Aldrich
1-Propanethiol, 99%
Sigma-Aldrich
Ethanethiol, ≥97%
Sigma-Aldrich
Sodium ethanethiolate, technical, ~90% (RT)
Sigma-Aldrich
Sodium ethanethiolate, technical grade, 80%