Merck
  • Transcriptional and posttranscriptional regulation of CYP1A1 by vanadium in human hepatoma HepG2 cells.
Transcriptional and posttranscriptional regulation of CYP1A1 by vanadium in human hepatoma HepG2 cells.
Cell biology and toxicology (2010-03-12)
Ghada Abdelhamid, Anwar Anwar-Mohamed, Osama A Badary, Adel A Moustafa, Ayman O S El-Kadi
ABSTRACT
We recently demonstrated that V(5+) downregulates 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD)-mediated induction of Cyp1a1 mRNA, protein, and catalytic activity levels in Hepa 1c1c7 cells through transcriptional mechanism. Therefore, it is important to investigate whether similar changes occur in humans. For this purpose, we examined the effect of V(5+) (as ammonium metavanadate, NH(4)VO(3)) on the expression of aryl hydrocarbon receptor (AhR)-regulated gene; cytochrome P450 1A1 (CYP1A1) at each step of the AhR signal transduction pathway in human hepatoma HepG2 cells. Our results show a significant reduction in TCDD-mediated induction of CYP1A1 mRNA, protein, and activity levels after V(5+) treatment in a dose-dependent manner. Investigating the effect of co-exposure to V(5+) and TCDD at transcriptional levels revealed that V(5+) significantly inhibited TCDD-mediated induction of AhR-dependent luciferase reporter gene expression. Looking at the posttranscriptional level, V(5+) did not affect CYP1A1 mRNA stability, thus eliminating the possible role of V(5+) in modifying CYP1A1 gene expression through this mechanism. On the other hand, at the posttranslational level, V(5+) was able to significantly decrease CYP1A1 protein half-life contributing to the inconsistency between catalytic activity and transcriptional level. Importantly, we showed that V(5+) did not significantly alter the heme oxygenase-1 mRNA level, thus eliminating any possibility that V(5+) might have decreased CYP1A1 activity through affecting its heme content. This study demonstrates for the first time that V(5+) downregulates the expression of CYP1A1 at the transcriptional, posttranscriptional and posttranslational mechanisms in the human hepatoma HepG2 cells.
Sigma-Aldrich
Ammonium metavanadate, 99%
Sigma-Aldrich
Ammonium metavanadate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium metavanadate, 99.95% trace metals basis
Sigma-Aldrich
Ammonium metavanadate, puriss. p.a., ACS reagent, ≥99.0% (RT)