- Parental dietary effect on embryological development response to toxicants with the sea urchin Arbacia punctulata.
Parental dietary effect on embryological development response to toxicants with the sea urchin Arbacia punctulata.
Bulletin of environmental contamination and toxicology (2009-11-26)
R W Nelson, M Nipper, A Lawrence, S Watts
PMID19936580
ABSTRACT
The role of echinoid parental nutrition in early-life stage toxicity is not well understood. Arbacia punctulata were fed either a fresh diet consisting of organic lettuce and carrots or a dry feed. Embryos from parents fed the dry feed exhibited lower sensitivity to copper, whereas the opposite occurred with 1,3,5-trinitrobenzene and sodium dodecyl sulfate (SDS). EC(50) values for the dry and fresh feed treatments, respectively, were 41.0 and 29.9 microg/L for copper, 0.5 and 1.8 mg/L for 1,3,5-trinitrobenzene, and 3.5 and 5.6 mg/L for SDS. The data suggests that nutritional standardization for sea urchins in ecotoxicological laboratories needs to be addressed and further investigated.