Merck
  • Contrasting synergistic anion effects in vanadium(V) binding to nicatransferrin versus human serum transferrin.
Contrasting synergistic anion effects in vanadium(V) binding to nicatransferrin versus human serum transferrin.
Biochemistry (2009-11-20)
Jean P Gaffney, Ann M Valentine
ABSTRACT
Some ascidians sequester vanadium and other metal ions that are bound and transported in higher organisms by transferrin. The ascidian Ciona intestinalis has a monolobal transferrin (nicatransferrin) in its plasma. The binding of vanadium(V) to nicatransferrin was investigated by using isothermal titration calorimetry and UV-vis spectroscopy, in the presence and absence of NaHCO(3), and was compared with human serum transferrin. Nicatransferrin and serum transferrin bind V(V) with similar strengths [K = (2.0 +/- 0.6) x 10(5) M(-1) for nicatransferrin]; however, nicatransferrin requires a synergistic anion for V(V) binding, whereas serum transferrin does not. Spectroscopy supports a different kind of coordination site.
Sigma-Aldrich
Ammonium metavanadate, 99.95% trace metals basis
Sigma-Aldrich
Ammonium metavanadate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Ammonium metavanadate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium metavanadate, 99%