Merck
  • Extracellular thiol-assisted selenium uptake dependent on the x(c)- cystine transporter explains the cancer-specific cytotoxicity of selenite.
Extracellular thiol-assisted selenium uptake dependent on the x(c)- cystine transporter explains the cancer-specific cytotoxicity of selenite.
Proceedings of the National Academy of Sciences of the United States of America (2009-06-25)
Eric Olm, Aristi P Fernandes, Christina Hebert, Anna-Klara Rundlöf, Erik H Larsen, Olof Danielsson, Mikael Björnstedt
ABSTRACT
The selenium salt selenite (SeO(3)(2-)) is cytotoxic in low to moderate concentrations, with a remarkable specificity for cancer cells resistant to conventional chemotherapy. Our data show that selenium uptake and accumulation, rather than intracellular events, are crucial to the specific selenite cytotoxicity observed in resistant cancer cells. We show that selenium uptake depends on extracellular reduction, and that the extracellular environment is a key factor specific to selenite cytotoxicity. The extracellular reduction is mediated by cysteine, and the efficacy is determined by the uptake of cystine by the x(c)(-) antiporter and secretion of cysteine by multidrug resistance proteins, both of which are frequently overexpressed by resistant cancer cells. This mechanism provides molecular evidence for the existence of an inverse relationship between resistance to conventional chemotherapy and sensitivity to selenite cytotoxicity, and highlights the great therapeutic potential in treating multidrug-resistant cancer.
Sigma-Aldrich
Seleno-L-cystine, 95%
Sigma-Aldrich
Seleno-DL-cystine, powder