Merck
  • Synergetic bitherapy in mice with xenografts of human prostate cancer using a methional mimic (METLICO) and an aldehyde dehydrogenase 3 inhibitor (MATE): systemic intraperitoneal (IP) and targeted intra-tumoral (IT) administration.
Synergetic bitherapy in mice with xenografts of human prostate cancer using a methional mimic (METLICO) and an aldehyde dehydrogenase 3 inhibitor (MATE): systemic intraperitoneal (IP) and targeted intra-tumoral (IT) administration.
Current medicinal chemistry (2009-04-10)
E Hiltbrand, G Fournet, J-P Giliberto, M-J Paret, J Chantepie, D Morel, J Goré, U Reichert, H Mehier, M-E Rochedix, G Quash
ABSTRACT
An intraperitoneal (IP) monotherapy in nu/nu mice with subcutaneous xenografts of a human prostate epithelial cancer cell line:DU145 was undertaken with an aldehyde dehydrogenase 3 inhibitor MATE, that is a potent apoptogen on (DU145) in culture but not on their human prostate epithelial normal counterparts [13] . Tumour growth was slowed down but treatment had to be done 5days/week. To try to potentiate the action of MATE in vivo, a bitherapy was undertaken based on the synergetic apoptotic effect that had been observed previously in culture on DU145 treated with a methional mimic METLICO and DIMATE, an inhibitor of ALDH1 and ALDH3 [19]. The bitherapy with METLICO/MATE administered IP was as effective as the monotherapy with MATE alone by IP, but at a 2-fold lower dose of MATE and at a dose of METLICO that had no growth-inhibitory effect as a monotherapy . Hence there was definite synergism with bitherapy. To try to increase the efficacy of bitherapy, it was administered by the intra-tumoral (IT) route using the recently developed 20-bars-pressurized microinjection system from CERMA [16, 17]. IT administration of the bitherapy was indeed more effective than that by IP as regards tumour volumes are concerned. Histopathological analysis of IT-treated tumours confirmed that there were many necrotized zones but intact cells were still present. Approaches for treating a wider zone of tumour tissue by IT-bitherapy are discussed.
Sigma-Aldrich
3-(Methylthio)propionaldehyde, 96%
Sigma-Aldrich
Methional, natural, ≥98%, FG
Sigma-Aldrich
Methional, ≥97%, FG