MilliporeSigma
  • Catalyst-free tandem aldol condensation/Michael addition of 1,3-cyclohexanediones with enolizable aldehydes.
Catalyst-free tandem aldol condensation/Michael addition of 1,3-cyclohexanediones with enolizable aldehydes.
Bioorganic & medicinal chemistry letters (2009-03-31)
Kerstin Rohr, Rainer Mahrwald
ABSTRACT
An efficient tandem aldol condensation/Michael addition process of unactivated aldehydes and 1,3-cyclohexanedione is described. This transformation proceeds without any catalyst at room temperature with high isolated yields. By a fine-tuning of reaction conditions an access to both the aldol condensation/Michael addition products or to the dehydrated cyclized 9-substituted 1,8-dioxo-xanthenes is given.
Sigma-Aldrich
1,3-Cyclohexanedione, 97%