Merck
  • 1-Nitropyrene efficiently induces mitotic recombination in Saccharomyces cerevisiae.
1-Nitropyrene efficiently induces mitotic recombination in Saccharomyces cerevisiae.
Journal of radiation research (2008-09-09)
Lalla Rajaa Rhenimi, Naglaa Fathi Abu-Nasr, Kazuo Yamamoto
ABSTRACT
Nitropyrene, a mutagenic and carcinogenic component of diesel exhaust, has been shown to be a potent bacterial and mammalian mutagen. There is, however, some controversy regarding the genotoxic effects of 1-nitropyrene towards yeast. To obtain insights into the mechanisms of 1-nitropyrene-induced mutations in Saccharomyces cerevisiae, we have attempted to characterize the genetic alterations that inactivate the endogenous CAN1 gene either in haploid cells or in heterozygous diploid cells. 1-Nitropyrene, without any activation treatment, showed a substantial toxic effect until 500 microM. The mutation frequency in haploid cells treated with 500 microM of 1-nitropyrene was 1.59 x 10(-5), which is 15-fold higher than the control value. Sequencing of mutants indicated that both frameshifts and base substitutions were increased. In diploid cells treated with 500 microM of 1-nitropyrene, the frequency with which can1Delta::LEU2/can1Delta::LEU2 was converted from CAN1/can1Delta::LEU2, a phenotypic change from a canavanine-sensitive to canavanine-resistant form, was 8.59 x 10(-4), which is 9.15-fold higher than the spontaneous level. More than 99% of the 1-nitropyrene-induced mutations in canavanine-resistant diploid cells constituted a gene conversion or crossover. Chromosome loss was not increased after treatment with 1-nitropyrene. These results suggest that 1-nitropyrene is an agent that efficiently induces point mutations, gene conversion, and crossover, but not chromosome loss, in S. cerevisiae.
Sigma-Aldrich
1-Nitropyrene, ≥95%
1-Nitropyrene, BCR®, certified reference material