Merck
  • A highly sensitive fluorogenic probe for cytochrome P450 activity in live cells.
A highly sensitive fluorogenic probe for cytochrome P450 activity in live cells.
Bioorganic & medicinal chemistry letters (2008-07-04)
Melissa M Yatzeck, Luke D Lavis, Tzu-Yuan Chao, Sunil S Chandran, Ronald T Raines
ABSTRACT
A derivative of rhodamine 110 has been designed and assessed as a probe for cytochrome P450 activity. This probe is the first to utilize a 'trimethyl lock' that is triggered by cleavage of an ether bond. In vitro, fluorescence was manifested by the CYP1A1 isozyme with k(cat)/K(M)=8.8x10(3)M(-1)s(-1) and K(M)=0.09microM. In cellulo, the probe revealed the induction of cytochrome P450 activity by the carcinogen 2,3,7,8-tetrachlorodibenzo-p-dioxin, and its repression by the chemoprotectant resveratrol.
Sigma-Aldrich
Rhodamine 110 chloride, Dye content ≥75 %
Sigma-Aldrich
Rhodamine 110 chloride, suitable for fluorescence, BioReagent, ≥99.0% (UV)