Merck
  • Porphyrin complexes containing coordinated BOB groups: synthesis, chemical reactivity and the structure of [BOB(tpClpp)]2+.
Porphyrin complexes containing coordinated BOB groups: synthesis, chemical reactivity and the structure of [BOB(tpClpp)]2+.
Dalton transactions (Cambridge, England : 2003) (2008-03-13)
Warwick J Belcher, Michael C Hodgson, Kenji Sumida, Ana Torvisco, Karin Ruhlandt-Senge, D C Ware, P D W Boyd, P J Brothers
ABSTRACT
The reactions of boron halides with free base porphyrins under conditions where partial hydrolysis of the boron halides can occur give diboron porphyrin complexes containing BOB moieties in which each boron is bonded to two porphyrin nitrogen atoms. BF(3).OEt(2) with H(2)(por) gives B(2)OF(2)(por) (por = tpp, ttp, tpClpp, oep) which has an asymmetric structure in which one boron lies in the porphyrin plane (B(ip)) while the other lies above it (B(oop)). BCl(3).MeCN with H(2)(por) gives B(2)O(2)(BCl(3))(2)(por) which contains a four-membered B(2)O(2) ring and is stable only in the presence of excess BCl(3). BBr(3) with Li(2)(tpClpp) gives the dicationic complex [B(2)O(tpClpp)](2+) as its [BBr(4)](-) salt, and is the first example of a boron porphyrin containing three-coordinate boron to be structurally characterised. B(2)O(2)(BCl(3))(2)(por) can be chromatographed on basic alumina to give the hydroxyboron complex B(2)O(OH)(2)(por), which is deduced from its NMR spectra and DFT calculations to have a structure analogous to B(2)OF(2)(por). The OH protons are shifted upfield to near delta -4 (B(oop)-OH) and -10 (B(ip)-OH) by the diamagnetic porphyrin ring current. The reaction of either B(2)O(2)(BCl(3))(2)(por) or B(2)O(OH)(2)(por) (por = ttp, tpClpp) with alcohols (ROH, R = Et, 4-C(6)H(4)CH(3)) gives B(2)O(OR)(2)(por), which can in turn be converted to B(2)O(OR)(OH)(por) by repeated chromatography. The reaction of PhBCl(2) with H(2)(por) (por = ttp, tpClpp) gives B(2)O(Ph)(OH)(por) which has been characterised by spectroscopy in concert with DFT calculations. It is a further example of the B(2)OF(2)(por) structural type, in which the phenyl group is coordinated to the out-of-plane boron and the OH group to the in-plane boron, as are its derivatives B(2)O(Ph)(X)(tpClpp) (X = F, OEt). Steric drivers for the facile hydrolysis of haloboron porphyrins relative to their dipyrromethene and expanded porphyrin counterparts are discussed.
Sigma-Aldrich
Boron trichloride solution, 1.0 M in toluene
Sigma-Aldrich
Boron trichloride solution, 1.0 M in heptane
Sigma-Aldrich
Boron trichloride solution, 1.0 M in p-xylene
Sigma-Aldrich
Boron trichloride solution, 1.0 M in methylene chloride