Merck
  • Synthesis of trans-fused tetrahydrooxepins: stereoselective allylation of sulfur or fluoro-substituted tetrahydrooxepins.
Synthesis of trans-fused tetrahydrooxepins: stereoselective allylation of sulfur or fluoro-substituted tetrahydrooxepins.
Carbohydrate research (2007-12-11)
Shoji Kobayashi, Makiko Hori, Masahiro Hirama
ABSTRACT
An efficient route to the trans-fused tetrahydrooxepin corresponding to the E ring of ciguatoxin was developed. Wide screening of allylation reactions of sulfur or fluoro-substituted tetrahydrooxepin revealed that the optimum method for obtaining the beta-allylation product selectively was the use of a combination of allyltrimethylsilane and TiCl(4) with 6-fluoro-7-hydroxytetrahydrooxepin.
Sigma-Aldrich
Allyltrimethylsilane, 98%