Merck
  • Dipolar cycloaddition of ethyl isocyanoacetate to 3-chloro-2-(methylthio)/2-(methylsulfonyl)quinoxalines: highly regio- and chemoselective synthesis of substituted imidazo[1,5-a]quinoxaline-3-carboxylates.
Dipolar cycloaddition of ethyl isocyanoacetate to 3-chloro-2-(methylthio)/2-(methylsulfonyl)quinoxalines: highly regio- and chemoselective synthesis of substituted imidazo[1,5-a]quinoxaline-3-carboxylates.
The Journal of organic chemistry (2007-06-02)
G S M Sundaram, B Singh, C Venkatesh, H Ila, H Junjappa
ABSTRACT
An efficient route for regio- and chemoselective synthesis of substituted 3-(carboethoxy)imidazo[1,5-a]quinoxalines and novel diimidazo[1,5-a:5',1'-c]quinoxalines via base-induced cycloaddition of ethyl isocyanoacetate to unsymmetrically substituted 3-chloro-2-(methylthio)/2-(methylsulfonyl)quinoxalines has been reported.
Sigma-Aldrich
Ethyl isocyanate, 98%