- [A synthesis of cinnamoyloxyisoflavones].
[A synthesis of cinnamoyloxyisoflavones].
Bioorganicheskaia khimiia (2006-08-17)
A Aĭtmambetov, A M Tokhtybaeva, R T Tlegenov, L Tsao
PMID16909870
ABSTRACT
Interaction of 7-hydroxyisoflavonones with cinnamoyl chloride results in cinnamoyloxyisoflavonones.