Merck
  • Unusual role of solvents in the syntheses of {Fe-NO}6,7 nitrosyls derived from a ligand with carboxamido nitrogen and thiolato sulfur donors.
Unusual role of solvents in the syntheses of {Fe-NO}6,7 nitrosyls derived from a ligand with carboxamido nitrogen and thiolato sulfur donors.
Inorganic chemistry (2005-09-27)
Todd C Harrop, Marilyn M Olmstead, Pradip K Mascharak
ABSTRACT
Reaction of excess NO with the S = 3/2 Fe(III) complex (Et4N)2[Fe(PhPepS)(Cl)] (1) in protic solvents such as MeOH affords the {Fe-NO}(7) nitrosyl (Et(4)N)(2)[Fe(PhPepS)(NO)] (2). This distorted square-pyramidal S = 1/2 complex, a product of reductive nitrosylation, is the first example of an {Fe-NO}7 nitrosyl with carboxamido-N and thiolato-S coordination. When the same reaction is performed in aprotic solvents such as MeCN and DMF, the product is a dimeric diamagnetic {Fe-NO}6 complex, (Et4N)2-[{Fe(PhPepS)(NO)}2] (3). Both electrochemical and chemical oxidation of 2 leads to the formation of 3 via a transient five-coordinate {Fe-NO}6 intermediate. The oxidation is NO-centered. The ligand frame is not attacked by excess NO in these reactions.
Sigma-Aldrich
Iron(III) nitrate nonahydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Iron(III) nitrate nonahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Iron(III) nitrate nonahydrate, ≥99.999% trace metals basis
Sigma-Aldrich
Iron(III) nitrate nonahydrate, ACS reagent, ≥98%