Merck
  • Determination of furaneol (4-hydroxy-2,5-dimethyl-3(2H)-furanone) in some wines from Italian native grapes by Gas-Chromatography-SIM/MASS spectrometry.
Determination of furaneol (4-hydroxy-2,5-dimethyl-3(2H)-furanone) in some wines from Italian native grapes by Gas-Chromatography-SIM/MASS spectrometry.
Annali di chimica (2005-09-03)
Alessandro Genovese, Paola Piombino, Maria Tiziana Lisanti, Luigi Moio
ABSTRACT
Gas Chromatography-Mass Spectrometry (GC-MS) analysis by Selective Ion Monitoring (SIM) was applied to quantify 4-Hydroxy-2,5-dimethyl-3(2H)-furanone (HDMF) in both red and white wines obtained from some Italian cultivar of Vitis vinifera. Wines were extracted by liquid-liquid extraction performed with 1,1,2-trichlorotrifluoroethane (Freon 113). The ion m/z 128 was used for quantification while the ion m/z 129 as qualifier. Precision, linearity and accuracy of the method resulted satisfactory. Results showed a significant variation in the concentration of furaneol in wine with grape variety. Generally, HDMF concentrations in white wines were lower than in red wines. Among white wines, Chardonnay resulted characterized by the highest concentration of HDMF. Among red wines the highest concentrations of HDMF were detected in Primitivo and Refosco varieties.
Sigma-Aldrich
4-Hydroxy-2,5-dimethyl-3(2H)-furanone, ≥99.0% (GC)
Supelco
4-Hydroxy-2,5-dimethyl-3(2H)-furanone, analytical standard
Sigma-Aldrich
4-Hydroxy-2,5-dimethyl-3(2H)-furanone, ≥98%, FCC, FG
Sigma-Aldrich
4-Hydroxy-2,5-dimethyl-3(2H)-furanone, natural, ≥98%, FG