Merck
  • Comparison between acetic acid and landfill leachates for the leaching of Pb(II), Cd(II), As(V), and Cr(VI) from cementitious wastes.
Comparison between acetic acid and landfill leachates for the leaching of Pb(II), Cd(II), As(V), and Cr(VI) from cementitious wastes.
Environmental science & technology (2004-08-10)
Cheryl E Halim, Jason A Scott, Helena Natawardaya, Rose Amal, Donia Beydoun, Gary Low
ABSTRACT
The Toxicity Characteristic Leaching Procedure (TCLP) has been widely used to characterize the suitability of solid wastes for disposal in landfills. However, the widespread application of this test for the assessment of wastes disposed in different landfill types is often questionable. This paper investigates the leaching profiles of cement-stabilized heavy metal ions, namely, Pb (II), Cd (II), As(V), and Cr(VI), using acetic acid and leachates from municipal and nonputrescible Australian landfill sites. The leaching profiles of Pb, Cd, As, and Cr using acetic acid were found to be similar to the nonputrescible landfill leachate and differed markedly from the municipal solid waste (MSW) leachate. The additional presence of high amounts of organic and inorganic compounds in the municipal landfill leachate influenced the leaching profiles of these metal ions as compared to the acetic acid and the nonputrescible systems. It is postulated that the organic compounds present in the municipal landfill leachate formed complexes with the Pb and Cd, increasing the mobility of these ions. Moreover, the organic compounds in the municipal landfill leachate induced a reducing environment in the leachate, causing the reduction of Cr(VI) to Cr(III). It was also found that the presence of carbonates in the municipal landfill leachate affected the stability of calcium arsenate, with the carbonate competing with arsenate for calcium at high pH, forcing arsenate into the solution.
Sigma-Aldrich
Sodium chromate, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chromate tetrahydrate, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cadmium nitrate tetrahydrate, 99.997% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cadmium nitrate tetrahydrate, 98%