Merck
  • Stage-specific surface chemicals of Plodia interpunctella: 2-acyl-1,3-cyclohexanediones from larval mandibular glands serve as cuticular lipids.
Stage-specific surface chemicals of Plodia interpunctella: 2-acyl-1,3-cyclohexanediones from larval mandibular glands serve as cuticular lipids.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology (2004-06-15)
Ralph W Howard, James E Baker
ABSTRACT
Cuticular lipid compositions of all life stages of the stored product moth Plodia interpunctella have been determined. Eggs and adults of P. interpunctella have cuticular lipids consisting solely of hydrocarbons. The composition of eggs and adult females is qualitatively nearly identical with ca. 86 hydrocarbons (11 n-alkanes, 39 monomethyl alkanes, 19 dimethyl alkanes, 11 trimethyl alkanes and 6 monoenes) except females lack the 2-methyl alkanes found in eggs. Adult males have a hydrocarbon composition qualitatively nearly identical to females with the exception that they lack the monoenes. Larval and pupal cuticular lipids are dominated by a mixture of ca. 20 previously described 2-acyl-1,3-cyclohexanediones, with only minute amounts of n-alkanes on the larvae and pupae. The 2-acyl-1,3-cyclohexanediones are continuously secreted onto their silk webbing and food particles by the paired mandibular glands found in all larvae. Extracts from dissected mandibular glands have a qualitatively identical composition to larval cuticular extracts. The pupal stage (which does not have mandibular glands) is enclosed in a silk cocoon also coated with 2-acyl-1,3-cyclohexanediones laid down while the wandering stage larvae spin the cocoon. The 2-acyl-1,3-cyclohexanediones have physical properties which closely mimic those of cuticular hydrocarbons, including melting point and boiling point range and hydrophobicity. This is the first report of an insect with a life stage that does not use conventional cuticular lipids for conservation of water.
Sigma-Aldrich
1,3-Cyclohexanedione, 97%