Merck
  • Solubilization of (+)-limonene by anionic/cationic mixed surfactant systems.
Solubilization of (+)-limonene by anionic/cationic mixed surfactant systems.
Drug development and industrial pharmacy (2003-05-14)
Yoshihiro Saito, Kaname Hashizaki, Hiroyuki Taguchi, Naotake Ogawa
ABSTRACT
The combined effects of sodium n-octyl sulfate (SOS) and cetyltrimethylammonium bromide (CTAB) on the solubilization of (+)-limonene in aqueous solution were studied using a headspace gas chromatography technique. The findings showed the mixing of SOS and CTAB resulted in positive synergistic effects on the solubility of (+)-limonene. The positive synergistic effects are explained from the perspective of the phase behavior of this mixed surfactant system.
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ≥98%
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, BioXtra